Diethyl propylmalonate ≥98.0% (by GC)

Supplier: TCI America

D4708-25ML D4708-5ML
TCD4708-25MLEA 67.94 CAD
TCD4708-25ML TCD4708-5ML
Diethyl propylmalonate ≥98.0% (by GC)
Diethyl propylmalonate
Formula: C₁₀H₁₈O₄
Boiling Pt: 222°C
Flash Pt: 92°C
MDL Number: MFCD00009168
CAS Number: 2163-48-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR