4-Butoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B4727-1G B4727-5G
TCB4727-1GEA 55.3 CAD
TCB4727-1G TCB4727-5G
4-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Butoxyphenylboronic acid
Formula: C₁₀H₁₅BO₃
MDL Number: MFCD03427054
CAS Number: 105365-51-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR