2-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
B4700-1G
TCB4700-1GEA
105.01
CAD
TCB4700-1G
2-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Butoxyphenylboronic acid
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...