5-Methylthiophene-2-carboxaldehyde ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde

M0911-25G M0911-5G
TCM0911-25GEA 115.34 CAD
TCM0911-25G TCM0911-5G
5-Methylthiophene-2-carboxaldehyde ≥97.0% (by GC)
5-Methylthiophene-2-carboxaldehyde
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 205°C
Flash Pt: 87°C
MDL Number: MFCD00005434
CAS Number: 13679-70-4
EINECS: 237-178-6

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark purple clear liquid to cloudy liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR