Coumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Oxo-2H-chromene-3-carboxylic acid

C3129-25G C3129-5G
TCC3129-25GEA 426.6 CAD
TCC3129-25G TCC3129-5G
Coumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Coumarin-3-carboxylic acid
Formula: C₁₀H₆O₄
MW: 190.16 g/mol
MDL Number: MFCD00006852
CAS Number: 531-81-7
EINECS: 208-518-0
UN: 2811
ADR: 6.1 III
Merck Index: 14,02563

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR