1-Butyl-1-methylpyrrolidiniumdicyanamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B5453-25G B5453-5G
76219-614EA 703.1 CAD
76219-614 76219-616
1-Butyl-1-methylpyrrolidiniumdicyanamide ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidinium dicyanamide
Formula: C₁₁H₂₀N₄
MDL Number: MFCD08458923
CAS Number: 370865-80-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR