1-Butyl-1-methylpyrrolidiniumtrifluormethansulfonate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Butyl-1-methylpyrrolidin-1-ium trifluoromethanesulphonate, 1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate, 1-Butyl-1-methylpyrrolidinium triflate, 1-Butyl-1-methylpyrrolidinium trifluoromethanesulphonate, 1-Butyl-1-methylpyrrolidin-1-ium trifluoromethanesulfonate
B5568-25G
B5568-5G
76218-844EA
451.88
CAD
76218-844
76218-846
1-Butyl-1-methylpyrrolidiniumtrifluormethansulfonate ≥98.0% (by total nitrogen basis)
1-Butyl-1-methylpyrrolidinium trifluormethansulphonate
Formula:
C₁₀H₂₀F₃NO₃S MW: 291.33 g/mol Melting Pt: 4°C |
MDL Number:
MFCD08458925 CAS Number: 367522-96-1 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...