Methyl-6-fluoronicotinate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate
M2724-1G
M2724-5G
TCM2724-1GEA
135.56
CAD
TCM2724-1G
TCM2724-5G
Methyl-6-fluoronicotinate ≥98.0% (by GC)
Methyl 6-fluoronicotinate
Formula:
C₇H₆FNO₂ MW: 155.13 |
MDL Number:
MFCD03095098 CAS Number: 1427-06-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...