Methyl-6-bromonicotinate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2682-1G
M2682-5G
TCM2682-1GEA
39.5
CAD
TCM2682-1G
TCM2682-5G
Methyl-6-bromonicotinate ≥98.0% (by GC)
Methyl 6-bromonicotinate
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD04972371 CAS Number: 26218-78-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...