4-Nitrophenylarsonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

N0964-1G N0964-200MG
TCN0964-1GEA 808.33 CAD
TCN0964-1G TCN0964-200MG
4-Nitrophenylarsonic acid ≥98.0% (by HPLC, titration analysis)
4-Nitrophenylarsonic acid
Formula: C₆H₆AsNO₅
Melting Pt: 300°C
MDL Number: MFCD00024575
CAS Number: 98-72-6
EINECS: 202-695-8
UN: 3465
ADR: 6.1,III
Merck Index: 14,06566

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR