Ethylenediammonium diiodide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

E1222-1G E1222-5G
TCE1222-1GEA 107.44 CAD
TCE1222-1G TCE1222-5G
Ethylenediammonium diiodide ≥98.0% (by total nitrogen and titration analysis)
Ethylenediammonium diiodide
Formula: C₂H₁₀I₂N₂
MDL Number: MFCD00035535
CAS Number: 5700-49-2
EINECS: 227-186-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR