2-Methoxy-4-nitrobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

M2656-1G M2656-5G
TCM2656-1GEA 128.93 CAD
TCM2656-1G TCM2656-5G
2-Methoxy-4-nitrobenzaldehyde ≥98.0% (by GC)
2-Methoxy-4-nitrobenzaldehyde
Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD02683052
CAS Number: 136507-15-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR