2,6-Dichloro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

D4707-1G D4707-5G
TCD4707-1GEA 77.74 CAD
TCD4707-1G TCD4707-5G
2,6-Dichloro-3-pyridineboronic acid (contains varying amounts of Anhydride)
2,6-Dichloro-3-pyridineboronic acid
Formula: C₅H₄BCl₂NO₂
Melting Pt: 150°C
MDL Number: MFCD03094989
CAS Number: 148493-34-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR