2,5-Dichloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
2,5-Dichloropyridine-4-boronic acid, 2,5-Dichloro-4-pyridylboronic acid
D5082-1G
D5082-5G
76219-840EA
148.52
CAD
76219-840
76219-842
2,5-Dichloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
2,5-Dichloro-4-pyridineboronic acid
Formula:
C₅H₄BCl₂NO₂ Melting Pt: 202°C |
MDL Number:
MFCD06798254 CAS Number: 847664-64-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 96.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...