3-Fluoro-2-formylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

F0975-1G F0975-200MG
TCF0975-1GEA 107.44 CAD
TCF0975-1G TCF0975-200MG
3-Fluoro-2-formylpyridine ≥98.0% (by GC)
3-Fluoro-2-formylpyridine
Formula: C₆H₄FNO
Boiling Pt: 112°C
MDL Number: MFCD07781234
CAS Number: 31224-43-8

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR