2-Acetamido-5-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

A2855-1G A2855-5G
TCA2855-1GEA 37.92 CAD
TCA2855-1G TCA2855-5G
2-Acetamido-5-nitropyridine ≥98.0% (by GC)
2-Acetamido-5-nitropyridine
Formula: C₇H₇N₃O₃
MDL Number: MFCD00833973
CAS Number: 5093-64-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR