N-Chlorosaccharin ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Chlorobenzo[d]isothiazol-3(2H)-one 1,1-dioxide, 2-Chloro-1,2-benzisothiazol-3(2H)-one 1,1-Dioxide

C1674-25G C1674-5G
TCC1674-25GEA 1090.2 CAD
TCC1674-25G TCC1674-5G
N-Chlorosaccharin ≥96.0% (by titrimetric analysis)
N-Chlorosaccharin
Formula: C₇H₄ClNO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00274277
CAS Number: 14070-51-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Iodometric Titration) min. 96.0 %
Melting point 144.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR