Ethyl-4-(trifluoromethyl)benzoate ≥98.0% (by GC)
Supplier: TCI America
E1330-25G
E1330-5G
TCE1330-25GEA
184.86
CAD
TCE1330-25G
TCE1330-5G
Ethyl-4-(trifluoromethyl)benzoate ≥98.0% (by GC)
Ethyl 4-(trifluoromethyl)benzoate
Formula:
C₁₀H₉F₃O₂ Boiling Pt: 80°C |
MDL Number:
MFCD00013563 CAS Number: 583-02-8 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...