Methyl-3-bromo-4-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 3-bromo-4-fluorobenzoate

M3018-1G M3018-5G
TCM3018-1GEA 47.4 CAD
TCM3018-1G TCM3018-5G
Methyl-3-bromo-4-fluorobenzoate ≥98.0% (by GC)
Methyl 3-bromo-4-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD04972915
CAS Number: 82702-31-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Acid Value max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR