4-Bromo-3-nitrobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

B4708-1G B4708-5G
TCB4708-1GEA 88.48 CAD
TCB4708-1G TCB4708-5G
4-Bromo-3-nitrobenzonitrile ≥98.0% (by GC)
4-Bromo-3-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00982006
CAS Number: 89642-49-9

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR