1-Hexylpyridinium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Hexylpyridin-1-ium hexafluorophosphate(V)
H1424-25G
H1424-5G
TCH1424-25GEA
1068.08
CAD
TCH1424-25G
TCH1424-5G
1-Hexylpyridinium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
1-Hexylpyridinium hexafluorophosphate
Formula:
C₁₁H₁₈F₆NP |
MDL Number:
MFCD03093298 CAS Number: 797789-00-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...