3-Chlorophthalic anhydride ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
C3489-25G
C3489-5G
TCC3489-25GEA
218.04
CAD
TCC3489-25G
TCC3489-5G
3-Chlorophthalic anhydride ≥98.0% (by GC)
3-Chlorophthalic anhydride
Formula:
C₈H₃ClO₃ Boiling Pt: 170°C |
MDL Number:
MFCD00023107 CAS Number: 117-21-5 EINECS: 204-179-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Morpholine Method) | min. 96.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...