Dimethyl-2-bromoterephthalate ≥98.0% (by GC)

Supplier: TCI America

D5003-25G D5003-5G
TCD5003-25GEA 782.1 CAD
TCD5003-25G TCD5003-5G
Dimethyl-2-bromoterephthalate ≥98.0% (by GC)
Dimethyl 2-bromoterephthalate
Formula: C₁₀H₉BrO₄
MDL Number: MFCD00060638
CAS Number: 18643-86-2
EINECS: 242-476-4

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 53.0 and le 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR