p-Toluic hydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2846-25G M2846-5G
TCM2846-25GEA 347.6 CAD
TCM2846-25G TCM2846-5G
p-Toluic hydrazide ≥98.0% (by GC, titration analysis)
p-Toluic hydrazide
Formula: C₈H₁₀N₂O
MDL Number: MFCD00007607
CAS Number: 3619-22-5
EINECS: 222-815-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR