Benzo[b]thiophene-3-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
B4774-1G
B4774-5G
TCB4774-1GEA
75.84
CAD
TCB4774-1G
TCB4774-5G
Benzo[b]thiophene-3-boronic acid (contains varying amounts of Anhydride)
Benzo[b]thiophene-3-boronic acid
Formula:
C₈H₇BO₂S Melting Pt: 230°C |
MDL Number:
MFCD01114669 CAS Number: 113893-08-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...