4-(4-Nitrophenyl)morpholine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N1124-25G N1124-5G
TCN1124-25GEA 99.54 CAD
TCN1124-25G TCN1124-5G
4-(4-Nitrophenyl)morpholine ≥98.0% (by GC, titration analysis)
4-(4-Nitrophenyl)morpholine
Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD00023317
CAS Number: 10389-51-2
EINECS: 233-851-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR