2-Morpholinoaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M2671-25G
M2671-5G
TCM2671-25GEA
259.12
CAD
TCM2671-25G
TCM2671-5G
2-Morpholinoaniline ≥98.0% (by GC, titration analysis)
2-Morpholinoaniline
Formula:
C₁₀H₁₄N₂O Boiling Pt: 120°C |
MDL Number:
MFCD00047408 CAS Number: 5585-33-1 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 94.0 to 98.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...