1-Butyl-2-thiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Butylthiourea
B4971-1G
B4971-5G
76219-476EA
150.1
CAD
76219-476
76219-478
1-Butyl-2-thiourea ≥98.0% (by total nitrogen basis)
1-Butyl-2-thiourea
Formula:
C₅H₁₂N₂S MW: 132.23 g/mol |
MDL Number:
MFCD00022173 CAS Number: 1516-32-1 EINECS: 216-165-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...