3'-Benzyloxyacetophenone ≥98.0% (by GC)

Supplier: TCI America

B4705-25G B4705-5G
TCB4705-25GEA 409.22 CAD
TCB4705-25G TCB4705-5G
3'-Benzyloxyacetophenone ≥98.0% (by GC)
3'-Benzyloxyacetophenone
Formula: C₁₅H₁₄O₂
Boiling Pt: 154°C
MDL Number: MFCD00026221
CAS Number: 34068-01-4
EINECS: 251-816-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR