2-Cyano-5-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3126-1G C3126-200MG
TCC3126-1GEA 200.66 CAD
TCC3126-1G TCC3126-200MG
2-Cyano-5-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
2-Cyano-5-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₂
MDL Number: MFCD06657825
CAS Number: 741709-63-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR