1-Benzyloxy-4-iodobenzene ≥98.0% (by GC)

Supplier: TCI America

B4692-25G B4692-5G
TCB4692-25GEA 472.74 CAD
TCB4692-25G TCB4692-5G
1-Benzyloxy-4-iodobenzene ≥98.0% (by GC)
1-Benzyloxy-4-iodobenzene
Formula: C₁₃H₁₁IO
MDL Number: MFCD00079704
CAS Number: 19578-68-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR