(2-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4737-1G B4737-5G
TCB4737-1GEA 126.4 CAD
TCB4737-1G TCB4737-5G
(2-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Bromo-4-methoxyphenyl)acetic acid
Formula: C₉H₉BrO₃
MDL Number: MFCD02664700
CAS Number: 66916-99-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR