3-Methylphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T3219-1G T3219-5G
TCT3219-1GEA 67.94 CAD
TCT3219-1G TCT3219-5G
3-Methylphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD05864303
CAS Number: 253342-48-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 34.0 to 38.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR