2-Methylphenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(o-tolyl)-1,3,2-dioxaborolane, 2-Methylphenylboronic acid, pinacol ester, o-Tolylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

T3195-1G T3195-5G
TCT3195-1GEA 82.16 CAD
TCT3195-1G TCT3195-5G
2-Methylphenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
2-Methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₂
MW: 218.1
MDL Number: MFCD06795658
CAS Number: 195062-59-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR