4-Cyanophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

T3350-1G T3350-5G
TCT3350-1GEA 112.18 CAD
TCT3350-1G TCT3350-5G
4-Cyanophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
MDL Number: MFCD03093897
CAS Number: 171364-82-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR