2-Fluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F0940-1G F0940-5G
TCF0940-1GEA 42.66 CAD
TCF0940-1G TCF0940-5G
2-Fluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MDL Number: MFCD02094728
CAS Number: 352303-67-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR