2-(4-Fluorophenyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

F0727-1G F0727-5G
76220-070EA 142.2 CAD
76220-070 76220-072
2-(4-Fluorophenyl)pyridine ≥98.0% (by GC)
2-(4-Fluorophenyl)pyridine
Formula: C₁₁H₈FN
Boiling Pt: 119°C
MDL Number: MFCD06201382
CAS Number: 58861-53-3
EINECS: 261-473-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR