3-Isothiocyanatopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

P2131-1G P2131-5G
TCP2131-1GEA 167.48 CAD
TCP2131-1G TCP2131-5G
3-Isothiocyanatopyridine ≥98.0% (by GC)
3-Isothiocyanatopyridine
Formula: C₆H₄N₂S
Boiling Pt: 65°C
Storage Temperature: Refrigerator
MDL Number: MFCD00052351
CAS Number: 17452-27-6

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR