3,5-Dinitrophenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Dinitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,5-Dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dinitrophenylboronic acid Pinacol ester

D5431-1G D5431-200MG
76219-980EA 377.62 CAD
76219-980 76219-994
3,5-Dinitrophenylboronic acid pinacol ester ≥98.0% (by GC)
3,5-Dinitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₆
MW: 294.07 g/mol
MDL Number: MFCD11504965
CAS Number: 428820-95-5

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR