2-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Nitrophenylboronic acid Pinacol ester, 2-Nitrobenzeneboronic acid pinacol ester, 2-Nitrophenylboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

N1006-1G N1006-5G
TCN1006-1GEA 96.38 CAD
TCN1006-1G TCN1006-5G
2-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BNO₄
MW: 249.07 g/mol
MDL Number: MFCD02179447
CAS Number: 190788-59-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR