3-Aminophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
T3028-1G
T3028-5G
TCT3028-1GEA
48.98
CAD
TCT3028-1G
TCT3028-5G
3-Aminophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Aminophenylboronic acid pinacol ester
Formula:
C₁₂H₁₈BNO₂ |
MDL Number:
MFCD03453668 CAS Number: 210907-84-9 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...