2-Aminophenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

T3543-1G T3543-5G
76220-678EA 90.06 CAD
76220-678 76220-680
2-Aminophenylboronic acid pinacol ester ≥98.0% (by GC)
2-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
Boiling Pt: 125°C
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR