2-(Benzyloxy)benzoic acid ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Benzyloxybenzoic acid
B4766-25G
B4766-5G
TCB4766-25GEA
515.08
CAD
TCB4766-25G
TCB4766-5G
2-(Benzyloxy)benzoic acid ≥98.0% (by GC)
2-(Benzyloxy)benzoic acid
Formula:
C₁₄H₁₂O₃ MW: 228.25 |
MDL Number:
MFCD00051940 CAS Number: 14389-86-7 UN: 3077 ADR: 9 III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...