4'-(Trifluoromethyl)biphenyl-2-carboxylic acid 98,0 GC, T
Supplier: TCI America
|
Warning
|
Synonyms:
4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
T3231-1G
T3231-5G
TCT3231-1GEA
56.88
CAD
TCT3231-1G
TCT3231-5G
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid 98,0 GC, T
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
Formula:
C₁₄H₉F₃O₂ MW: 266.22 |
MDL Number:
MFCD00075353 CAS Number: 84392-17-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...