1-Ethyl-3-methylimidazoliumthiocyanate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate

E0776-25G E0776-5G
76220-022EA 393.42 CAD
76220-022 76220-024
1-Ethyl-3-methylimidazoliumthiocyanate ≥98.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium thiocyanate
Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
Melting Pt: -6°C
MDL Number: MFCD06798182
CAS Number: 331717-63-6

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR