2,4-Dimethoxybenzaldehyde oxime ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
D4851-25G
D4851-5G
76219-724EA
289.14
CAD
76219-724
76219-726
2,4-Dimethoxybenzaldehyde oxime ≥95.0% (by GC)
2,4-Dimethoxybenzaldehyde oxime
Formula:
C₉H₁₁NO₃ |
MDL Number:
MFCD00082783 CAS Number: 31874-34-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...