4-Bromo-3-nitrobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5101-1G
B5101-5G
TCB5101-1GEA
41.08
CAD
TCB5101-1G
TCB5101-5G
4-Bromo-3-nitrobenzaldehyde ≥98.0% (by GC)
4-Bromo-3-nitrobenzaldehyde
Formula:
C₇H₄BrNO₃ MW: 230.02 g/mol |
MDL Number:
MFCD00204031 CAS Number: 163596-75-6 |
Specification Test Results
Appearance | White to Light red to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 107.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...