1-Butyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B5526-25G B5526-5G
76218-836EA 532.46 CAD
76218-836 76218-838
1-Butyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)
1-Butyl-3-methylimidazolium trifluoroacetate
Formula: C₁₀H₁₅F₃N₂O₂
MDL Number: MFCD08458921
CAS Number: 174899-94-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR