3-Bromoanthranilic acid ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5177-1G B5177-5G
TCB5177-1GEA 79 CAD
TCB5177-1G TCB5177-5G
3-Bromoanthranilic acid ≥98.0% (by HPLC)
3-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MDL Number: MFCD03618453
CAS Number: 20776-51-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR