6-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Fluoro-6-hydroxybenzaldehyde

F0988-1G F0988-5G
TCF0988-1GEA 234.79 CAD
TCF0988-1G TCF0988-5G
6-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
6-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01090996
CAS Number: 38226-10-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR